Merck
  • Impurity profiling of trinitrotoluene using vacuum-outlet gas chromatography-mass spectrometry.
Impurity profiling of trinitrotoluene using vacuum-outlet gas chromatography-mass spectrometry.
Journal of chromatography. A (2014-12-10)
Hanneke Brust, Sander Willemse, Tuoyu Zeng, Arian van Asten, Mattijs Koeberg, Antoine van der Heijden, Annabel Bolck, Peter Schoenmakers
ABSTRACT
In this work, a reliable and robust vacuum-outlet gas chromatography-mass spectrometry (GC-MS) method is introduced for the identification and quantification of impurities in trinitrotoluene (TNT). Vacuum-outlet GC-MS allows for short analysis times; the analysis of impurities in TNT was performed in 4min. This study shows that impurity profiling of TNT can be used to investigate relations between TNT samples encountered in forensic casework. A wide variety of TNT samples were analyzed with the developed method. Dinitrobenzene, dinitrotoluene, trinitrotoluene and amino-dinitrotoluene isomers were detected at very low levels (<1wt.%) by applying the MS in selected-ion monitoring (SIM) mode. Limits of detection ranged from 6ng/mL for 2,6-dinitrotoluene to 43ng/mL for 4-amino-2,6-dinitrotoluene. Major impurities in TNT were 2,4-dinitrotoluene and 2,3,4-trinitrotoluene. Impurity profiles based on seven compounds showed to be useful to TNT samples from different sources. Statistical analysis of these impurity profiles using likelihood ratios demonstrated the potential to investigate whether two questioned TNT samples encountered in forensic casework are from the same source.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Methyl decanoate, ≥99%, FG
Supelco
Methyl decanoate, analytical standard
Sigma-Aldrich
Methyl decanoate, 99%