Merck
  • Ex vivo bone morphogenetic protein 2 gene delivery using periodontal ligament stem cells for enhanced re-osseointegration in the regenerative treatment of peri-implantitis.
Ex vivo bone morphogenetic protein 2 gene delivery using periodontal ligament stem cells for enhanced re-osseointegration in the regenerative treatment of peri-implantitis.
Journal of biomedical materials research. Part A (2014-03-13)
Shin-Young Park, Kyoung-Hwa Kim, Eun-Hye Gwak, Sang-Hoon Rhee, Jeong-Cheol Lee, Seung-Yun Shin, Ki-Tae Koo, Yong-Moo Lee, Yang-Jo Seol
ABSTRACT
Peri-implantitis is a chronic inflammatory process with advanced bone loss and impaired healing potential. For peri-implantitis treatment, tissue engineering can be applied to enhance bone regeneration of peri-implant defects. This study aimed to evaluate ex vivo bone morphogenetic protein 2 (BMP2) gene delivery using canine periodontal ligament stem cells (PDLSCs) for regeneration of peri-implantitis defects. Canine PDLSCs were transduced with adenoviral vectors containing BMP2 (BMP2/PDLSCs). After peri-implantitis was induced by ligature placement in six beagle dogs, regenerative procedures were performed; hydroxyapatite (HA) particles and collagen gel with autologous canine PDLSCs (PDLSC group) or BMP2/PDLSCs (BMP/PDLSC group) or without cells (control group) were grafted into the defects and covered by an absorbable membrane. Three months later, the animals were sacrificed. In vitro, BMP2/PDLSCs showed similar levels of stem cell properties to PDLSCs, such as colony-forming efficiency and expression of MSC markers STRO-1 and CD 146. BMP2/PDLSCs produced BMP-2 until day 21 at a concentration of 4-8 ng/mL. In vivo, the BMP2/PDLSC group showed significantly more new bone formation and re-osseointegration in peri-implantitis defects compared to the other groups. In conclusion, ex vivo BMP2 gene delivery using PDLSCs enhanced new bone formation and re-osseointegration in peri-implantitis defects.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
USP
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Oxytetra Selective Supplement, suitable for microbiology