Merck
  • FRET detection of lymphocyte function-associated antigen-1 conformational extension.
FRET detection of lymphocyte function-associated antigen-1 conformational extension.
Molecular biology of the cell (2014-11-08)
Alexandre Chigaev, Yelena Smagley, Mark K Haynes, Oleg Ursu, Cristian G Bologa, Liliana Halip, Tudor Oprea, Anna Waller, Mark B Carter, Yinan Zhang, Wei Wang, Tione Buranda, Larry A Sklar
ABSTRACT
Lymphocyte function-associated antigen 1 (LFA-1, CD11a/CD18, αLβ2-integrin) and its ligands are essential for adhesion between T-cells and antigen-presenting cells, formation of the immunological synapse, and other immune cell interactions. LFA-1 function is regulated through conformational changes that include the modulation of ligand binding affinity and molecular extension. However, the relationship between molecular conformation and function is unclear. Here fluorescence resonance energy transfer (FRET) with new LFA-1-specific fluorescent probes showed that triggering of the pathway used for T-cell activation induced rapid unquenching of the FRET signal consistent with extension of the molecule. Analysis of the FRET quenching at rest revealed an unexpected result that can be interpreted as a previously unknown LFA-1 conformation.
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
2-(2-Aminoethyl)-1-methylpyrrolidine, 97%
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies