Merck
The cGAS-cGAMP-STING pathway of cytosolic DNA sensing and signaling.
Molecular cell (2014-04-29)
Xin Cai, Yu-Hsin Chiu, Zhijian J Chen
ABSTRACT
The innate immune system deploys a variety of sensors to detect signs of infection. Nucleic acids represent a major class of pathogen signatures that can trigger robust immune responses. The presence of DNA in the cytoplasm of mammalian cells is a danger signal that activates innate immune responses; however, how cytosolic DNA triggers these responses remained unclear until recently. In this review, we focus on the mechanism of DNA sensing by the newly discovered cGAS-cGAMP-STING pathway and highlight recent progress in dissecting the in vivo functions of this pathway in immune defense as well as autoimmunity.
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female