Merck
  • How cells sense extracellular matrix stiffness: a material's perspective.
How cells sense extracellular matrix stiffness: a material's perspective.
Current opinion in biotechnology (2013-04-25)
Britta Trappmann, Christopher S Chen
ABSTRACT
The mechanical properties of the extracellular matrix (ECM) in which cells reside have emerged as an important regulator of cell fate. While materials based on natural ECM have been used to implicate the role of substrate stiffness for cell fate decisions, it is difficult in these matrices to isolate mechanics from other structural parameters. In contrast, fully synthetic hydrogels offer independent control over physical and adhesive properties. New synthetic materials that also recreate the fibrous structural hierarchy of natural matrices are now being designed to study substrate mechanics in more complex ECMs. This perspective examines the ways in which new materials are being used to advance our understanding of how ECM stiffness impacts cell function.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor