Merck
  • A diastereoselective synthesis of 7α-nitromethyl steroid derivative and its use for an efficient synthesis of eplerenone.
A diastereoselective synthesis of 7α-nitromethyl steroid derivative and its use for an efficient synthesis of eplerenone.
Steroids (2012-05-17)
Bin Zhang, Hongli Chen, Huanyu Tang, Huijin Feng, Yuanchao Li
ABSTRACT
A novel and efficient method of stereoselectively introducing α-nitromethyl group to C-7 position of 11α-hydroxyl canrenone 4 was described. In addition, this method was successfully applied in a total synthesis of Eplerenone 8. The route was characteristic of simple operation, moderate reaction conditions with 5 steps and 55% total yield, at the same time, without any expensive or toxic reagent in use.
Product Description
Canrenone, European Pharmacopoeia (EP) Reference Standard