Merck
  • Development of an analytical method for 3-monochloropropane-1,2-diol in soy sauce using 4-heptanone as derivatizing agent.
Development of an analytical method for 3-monochloropropane-1,2-diol in soy sauce using 4-heptanone as derivatizing agent.
Food additives and contaminants (2004-06-16)
Fabian M Dayrit, Milady R Niñonuevo
ABSTRACT
3-Monochloro-1,2-propane diol is a suspected carcinogen found in hydrolysed vegetable protein products such as soy sauce. A method is described for the analysis of 3-monochloro-1,2-propane diol in soy sauce by gas chromatography-mass spectrometry at a concentration range of 1-5000 ng g-1 using 4-heptanone as the derivatizing ketone and 3-monochloro-1,2-propane diol-d5 as the internal standard. The limit of detection for the method in the soy sauce matrix was 0.48 ng g-1 and the limit of quantification was 1.2 ng g-1.
Sigma-Aldrich
4-Heptanone, 98%
Sigma-Aldrich
4-Heptanone, ≥97%, FG
Supelco
4-Heptanone, analytical standard