Merck
  • Fast nucleation and growth of ZIF-8 nanocrystals monitored by time-resolved in situ small-angle and wide-angle X-ray scattering.
Fast nucleation and growth of ZIF-8 nanocrystals monitored by time-resolved in situ small-angle and wide-angle X-ray scattering.
Angewandte Chemie (International ed. in English) (2011-07-13)
Janosch Cravillon, Christian A Schröder, Roman Nayuk, Jeremie Gummel, Klaus Huber, Michael Wiebcke
ABSTRACT
Prenucleation clusters: in situ synchrotron X-ray scattering with a one-second time resolution revealed the occurrence of nano-sized clusters during the nucleation and early growth of nanocrystals of a zeolitic imidazolate framework (ZIF). The complex crystallization process exhibits similarities with crystallization processes of zeolites from solution. Hmim= 2-methylimidazole.
Sigma-Aldrich
2-Methylimidazole, 99%