Merck
  • Palladium-catalyzed regioselective carbonylation of C-H bonds of N-alkyl anilines for synthesis of isatoic anhydrides.
Palladium-catalyzed regioselective carbonylation of C-H bonds of N-alkyl anilines for synthesis of isatoic anhydrides.
Journal of the American Chemical Society (2012-10-12)
Zheng-Hui Guan, Ming Chen, Zhi-Hui Ren
ABSTRACT
A Pd-catalyzed regioselective C-H bond carbonylation of N-alkyl anilines for the synthesis of isatoic anhydrides has been developed. The key Pd-catalyst intermediate has been isolated and characterized. This novel Pd-catalyzed carbonylation reaction tolerates a wide range of functional groups and is a reliable method for the rapid elaboration of readily available N-alkyl anilines into a variety of substituted isatoic anhydrides under mild conditions.
Sigma-Aldrich
Isatoic anhydride, 96%