Merck
  • [Headspace GC/MS analysis of residual vinyl chloride and vinylidene chloride in polyvinyl chloride and polyvinylidene chloride products].
[Headspace GC/MS analysis of residual vinyl chloride and vinylidene chloride in polyvinyl chloride and polyvinylidene chloride products].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2005-05-11)
Hiroyuki Ohno, Motoh Mutsuga, Yoko Kawamura, Masako Suzuki, Taiki Aoyama
ABSTRACT
A headspace GC/MS analysis method for the simultaneous determination of residual vinyl chloride (VC) and vinylidene chloride (VDC) in polyvinyl chloride (PVC) and polyvinylidene chloride (PVDC) products was developed. A test sample was swelled overnight with N,N-dimethylacetamide in a sealed vial. The vial was incubated for 1 hour at 90 degrees C, then the headspace gas was analyzed by GC/MS using a PLOT capillary column. The recoveries from spiked PVC and PVDC samples were 90.0-112.3% for VC and 85.2-108.3% for VDC. The determination limits were 0.01 microg/g for VC and 0.06/microg/g for VDC, respectively. By this method, VC was detected in two PVC water supply pipes at the levels of 0.61 and 0.01 microg/g. On the other hand, VC and VDC were not detected in any of the food container-packages or toys tested.
Product Description
Supelco
1,1-Dichloroethylene, analytical standard
Supelco
1,1-Dichloroethene solution, certified reference material, 1000 μg/mL in methanol