Merck
  • A new family of small molecules to probe the reactivation of mutant p53.
A new family of small molecules to probe the reactivation of mutant p53.
Journal of the American Chemical Society (2005-04-28)
Michael C Myers, Jinling Wang, Jaclyn A Iera, Jeong-Kyu Bang, Toshiaki Hara, Shin'ichi Saito, Gerard P Zambetti, Daniel H Appella
ABSTRACT
Cells that express mutant p53 derived from cancers are selectively killed by a new class of small organic molecules. The protein p53 is recognized as one of the most important guardians in the body that prevents tumor development. Mutant forms of p53 are present in approximately 50% of all human cancers. Molecules that selectively kill cells expressing mutant p53 could become important chemotherapeutic agents. Our research focuses on developing a synthetically accessible class of molecules that can be easily modified to examine structural activity relationships and mechanism of biological activity or to optimize for anticancer activity. In this communication, a new class of molecules that selectively arrests growth of cells expressing two forms of mutant p53 is described. Synthetic routes to these compounds are also presented.
Sigma-Aldrich
2′-Aminoacetophenone, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
2′-Aminoacetophenone, 98%
Pricing and availability is not currently available.