Merck
Uptake and reduction of alpha-lipoic acid by human erythrocytes.
Clinical biochemistry (2007-08-04)
James M May, Zhi-chao Qu, Deanna J Nelson
ABSTRACT
The reducing capacity of erythrocytes has been used clinically as to estimate resistance to oxidant stress. In this work we targeted the antioxidant capacity of pyridine nucleotide disulfide reductases of these cells by measuring their ability to reduce the disulfide alpha-lipoic acid. Erythrocyte reduction of alpha-lipoic acid and related disulfides was measured as reduction of 5,5'-dithiobis(2-nitrobenzoic acid) (DTNB) outside the cells. Lipoic acid-dependent DTNB reduction by human erythrocytes required d-glucose and consumed NADPH, but not NADH. This activity was inhibited by carmustine and phenylarsine oxide, as expected if alpha-lipoic acid is reduced by the glutathione and thioredoxin reductase systems. Reduction of hydroxyethyl disulfide, which provides an estimate of total erythrocyte disulfide reduction capacity, was similar to that of alpha-lipoic acid. Erythrocytes incubated with alpha-lipoic acid also reduced extracellular ferricyanide, although rates of dehydroascorbate reduction were several-fold greater, probably because intracellular GSH can recycle ascorbate but not alpha-lipoic acid in erythrocytes. These results show that alpha-lipoic acid-dependent DTNB reduction provides a simple method to selectively assess the capacity of pyridine nucleotide disulfide reductases of human erythrocytes. When coupled with other non-destructive assays, such as reduction of hydroxyethyl disulfide and ferricyanide, this assay provides a comprehensive approach to assessing erythrocyte reducing capacity in a variety of clinical conditions associated with oxidant stress.
Sigma-Aldrich
Thioredoxin from Escherichia coli, recombinant, expressed in E. coli, essentially salt-free, lyophilized powder, ≥3 units/mg protein
Sigma-Aldrich
Thioredoxin Reductase from rat liver, buffered aqueous glycerol solution, ≥100 units/mg protein (Bradford)