Merck
  • Axonal transport of Frizzled5 by Alcadein α-containing vesicles is associated with kinesin-1.
Axonal transport of Frizzled5 by Alcadein α-containing vesicles is associated with kinesin-1.
Molecular biology of the cell (2023-08-16)
Yuzuha Shiraki, Monet Mitsuma, Ritsuko Takada, Saori Hata, Akira Kitamura, Shinji Takada, Masataka Kinjo, Hidenori Taru, Ulrike C Müller, Tohru Yamamoto, Yuriko Sobu, Toshiharu Suzuki
ABSTRACT
Alcadein α (Alcα) and amyloid-β protein precursor (APP) are cargo receptors that associate vesicles with kinesin-1. These vesicles, which contain either Alcα or APP, transport various proteins/cargo molecules into axon nerve terminals. Here, we analyzed immune-isolated Alcα- and APP-containing vesicles of adult mouse brains with LC-MS/MS and identified proteins present in vesicles that contained either Alcα or APP. Among these proteins, Frizzled-5 (Fzd5), a Wnt receptor, was detected mainly in Alcα vesicles. Although colocalization ratios of Fzd5 with Alcα are low in the neurites of differentiating neurons by a low expression of Fzd5 in embryonic brains, the suppression of Alcα expression decreased the localization of Fzd5 in neurites of primary cultured neurons. Furthermore, Fzd5-EGFP expressed in primary cultured neurons was preferentially transported in axons with the transport velocities of Alcα vesicles. In synaptosomal fractions of adult-mice brains that express higher levels of Fzd5, the amount of Fzd5 and the phosphorylation level of calcium/calmodulin-dependent protein kinase-II were reduced in the Alcα-deficient mice. These results suggest that reduced transport of Fzd5 by Alcα-containing vesicles associated with kinesin-1 in axon terminals may impair the response to Wnt ligands in the noncanonical Ca2+-dependent signal transduction pathway at nerve terminals of mature neurons.
Sigma-Aldrich
Anti-VGluT1, from rabbit
Sigma-Aldrich
Anti-Actin, from rabbit, purified by affinity chromatography