Merck
  • Lipid levels correlate with neuronal and dopaminergic markers during the differentiation of SH-SY5Y cells.
Lipid levels correlate with neuronal and dopaminergic markers during the differentiation of SH-SY5Y cells.
Biochimica et biophysica acta. Molecular basis of disease (2024-05-16)
Frederik Ravnkilde Marlet, Sonia Sanz Muñoz, Nefeli Sotiraki, Jannik Nicklas Eliasen, Jakob Woessmann, Jan Weicher, Jesper Elmsted Dreier, Erwin M Schoof, Kristi A Kohlmeier, Kenji Maeda, Céline Galvagnion
ABSTRACT
Parkinson's Disease (PD) is characterised by the loss of dopaminergic neurons and the deposition of protein inclusions called Lewy Bodies (LBs). LBs are heterogeneous structures composed of protein and lipid molecules and their main constituent is the presynaptic protein α-synuclein. SH-SY5Y cells are neuroblastoma cells commonly used to model PD because they express dopaminergic markers and α-synuclein and they can be differentiated into neuronal cells using established protocols. Despite increasing evidence pointing towards a role of lipids in PD, limited knowledge is available on the lipidome of undifferentiated and differentiated SH-SY5Y cells. Using a combination of lipidomics, proteomics, morphological and electrophysiological measurements, we identified specific lipids, including sphingolipids, whose levels are affected by the differentiation of SH-SY5Y neuroblastoma cells and found that the levels of these lipids correlate with those of neuronal and dopaminergic markers. These results provide a quantitative characterisation of the changes in lipidome associated with the differentiation of SH-SY5Y cells into more neuronal and dopaminergic-like phenotype and serve as a basis for further characterisation of lipid disruptions in association with PD and its risk factors in this dopaminergic-like neuronal cell model.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Dopamine D3 Receptor Antibody, cytoplasmic domain, Chemicon®, from rabbit