Merck
  • Effects of hepatic mitochondrial pyruvate carrier deficiency on de novo lipogenesis and gluconeogenesis in mice.
Effects of hepatic mitochondrial pyruvate carrier deficiency on de novo lipogenesis and gluconeogenesis in mice.
iScience (2023-11-09)
Nicole K H Yiew, Stanislaw Deja, Daniel Ferguson, Kevin Cho, Chaowapong Jarasvaraparn, Miriam Jacome-Sosa, Andrew J Lutkewitte, Sandip Mukherjee, Xiaorong Fu, Jason M Singer, Gary J Patti, Shawn C Burgess, Brian N Finck
ABSTRACT
The liver coordinates the systemic response to nutrient deprivation and availability by producing glucose from gluconeogenesis during fasting and synthesizing lipids via de novo lipogenesis (DNL) when carbohydrates are abundant. Mitochondrial pyruvate metabolism is thought to play important roles in both gluconeogenesis and DNL. We examined the effects of hepatocyte-specific mitochondrial pyruvate carrier (MPC) deletion on the fasting-refeeding response. Rates of DNL during refeeding were impaired by hepatocyte MPC deletion, but this did not reduce intrahepatic lipid content. During fasting, glycerol is converted to glucose by two pathways; a direct cytosolic pathway and an indirect mitochondrial pathway requiring the MPC. Hepatocyte MPC deletion reduced the incorporation of 13C-glycerol into TCA cycle metabolites, but not into new glucose. Furthermore, suppression of glycerol and alanine metabolism did not affect glucose concentrations in fasted hepatocyte-specific MPC-deficient mice, suggesting multiple layers of redundancy in glycemic control in mice.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-acetyl CoA Carboxylase Antibody, clone 7D2.2, clone 7D2.2, from mouse
Sigma-Aldrich
Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody, Upstate®, from rabbit