Merck
  • The structure and spontaneous curvature of clathrin lattices at the plasma membrane.
The structure and spontaneous curvature of clathrin lattices at the plasma membrane.
Developmental cell (2021-04-07)
Kem A Sochacki, Bridgette L Heine, Gideon J Haber, John R Jimah, Bijeta Prasai, Marco A Alfonzo-Méndez, Aleah D Roberts, Agila Somasundaram, Jenny E Hinshaw, Justin W Taraska
ABSTRACT
Clathrin-mediated endocytosis is the primary pathway for receptor and cargo internalization in eukaryotic cells. It is characterized by a polyhedral clathrin lattice that coats budding membranes. The mechanism and control of lattice assembly, curvature, and vesicle formation at the plasma membrane has been a matter of long-standing debate. Here, we use platinum replica and cryoelectron microscopy and tomography to present a structural framework of the pathway. We determine the shape and size parameters common to clathrin-mediated endocytosis. We show that clathrin sites maintain a constant surface area during curvature across multiple cell lines. Flat clathrin is present in all cells and spontaneously curves into coated pits without additional energy sources or recruited factors. Finally, we attribute curvature generation to loosely connected and pentagon-containing flat lattices that can rapidly curve when a flattening force is released. Together, these data present a universal mechanistic model of clathrin-mediated endocytosis.
Sigma-Aldrich
Jasplakinolide, Jaspis johnstoni, InSolution, ≥90%, 1 mM in DMSO, mammablian cell mitosis inhibitor, potent inducer of actin polymerization and stabilization
Sigma-Aldrich
Latrunculin A, Latrunculia magnifica, Latrunculin A, CAS 76343-93-6, is a cell-permeable marine toxin that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (KD = 200 nM).
Sigma-Aldrich
Cilengitide trifluoroacetic acid salt, ≥95% (HPLC)