Merck
  • Glucuronoxylan-based quince seed hydrogel: A promising scaffold for tissue engineering applications.
Glucuronoxylan-based quince seed hydrogel: A promising scaffold for tissue engineering applications.
International journal of biological macromolecules (2021-03-25)
Meltem Guzelgulgen, Dilce Ozkendir-Inanc, Umit Hakan Yildiz, Ahu Arslan-Yildiz
ABSTRACT
Natural gums and mucilages from plant-derived polysaccharides are potential candidates for a tissue-engineering scaffold by their ability of gelation and biocompatibility. Herein, we utilized Glucuronoxylan-based quince seed hydrogel (QSH) as a scaffold for tissue engineering applications. Optimization of QSH gelation was conducted by varying QSH and crosslinker glutaraldehyde (GTA) concentrations. Structural characterization of QSH was done by Fourier Transform Infrared Spectroscopy (FTIR). Furthermore, morphological and mechanical investigation of QSH was performed by Scanning Electron Microscopy (SEM) and Atomic Force Microscopy (AFM). The protein adsorption test revealed the suitability of QSH for cell attachment. Biocompatibility of QSH was confirmed by culturing NIH-3T3 mouse fibroblast cells on it. Cell viability and proliferation results revealed that optimum parameters for cell viability were 2 mg mL-1 of QSH and 0.03 M GTA. SEM and DAPI staining results indicated the formation of spheroids with a diameter of approximately 300 μm. Furthermore, formation of extracellular matrix (ECM) microenvironment was confirmed with the Collagen Type-I staining. Here, it was demonstrated that the fabricated QSH is a promising scaffold for 3D cell culture and tissue engineering applications provided by its highly porous structure, remarkable swelling capacity and high biocompatibility.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Milli-Mark® Anti-Collagen Type I-FITC Antibody, clone 5D8-G9, clone 5D8-G9, Milli-Mark®, from mouse