Merck
  • Mitochondrial morphodynamics alteration induced by influenza virus infection as a new antiviral strategy.
Mitochondrial morphodynamics alteration induced by influenza virus infection as a new antiviral strategy.
PLoS pathogens (2021-02-18)
Irene Pila-Castellanos, Diana Molino, Joe McKellar, Laetitia Lines, Juliane Da Graca, Marine Tauziet, Laurent Chanteloup, Ivan Mikaelian, Laurène Meyniel-Schicklin, Patrice Codogno, Jacky Vonderscher, Cédric Delevoye, Olivier Moncorgé, Eric Meldrum, Caroline Goujon, Etienne Morel, Benoit de Chassey
ABSTRACT
Influenza virus infections are major public health threats due to their high rates of morbidity and mortality. Upon influenza virus entry, host cells experience modifications of endomembranes, including those used for virus trafficking and replication. Here we report that influenza virus infection modifies mitochondrial morphodynamics by promoting mitochondria elongation and altering endoplasmic reticulum-mitochondria tethering in host cells. Expression of the viral RNA recapitulates these modifications inside cells. Virus induced mitochondria hyper-elongation was promoted by fission associated protein DRP1 relocalization to the cytosol, enhancing a pro-fusion status. We show that altering mitochondrial hyper-fusion with Mito-C, a novel pro-fission compound, not only restores mitochondrial morphodynamics and endoplasmic reticulum-mitochondria contact sites but also dramatically reduces influenza replication. Finally, we demonstrate that the observed Mito-C antiviral property is directly connected with the innate immunity signaling RIG-I complex at mitochondria. Our data highlight the importance of a functional interchange between mitochondrial morphodynamics and innate immunity machineries in the context of influenza viral infection.
Sigma-Aldrich
2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Trypsin, TPCK-Treated
Roche
Cytotoxicity Detection Kit (LDH), suitable for protein quantification, suitable for cell analysis, detection, sufficient for ≤2,000 tests