Merck
  • Effect of high pressure processing on migration characteristics of polypropylene used in food contact materials.
Effect of high pressure processing on migration characteristics of polypropylene used in food contact materials.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2021-02-05)
Yoon S Song, John L Koontz, Rima O Juskelis, Eduardo Patazca, William Limm, Kun Zhao
ABSTRACT
The migration of small molecular mass organic compounds from polypropylene (PP) copolymer films into food simulants during and after high pressure processing (HPP) was studied. An overlapping temperature profile was developed to isolate the pressure effect of HPP (700 MPa, 71°C, 5 min) from equivalent thermal processing (TP) at atmospheric pressure (0.1 MPa). Chloroform, toluene, methyl salicylate, and phenylcyclohexane were chosen as surrogate compounds, and were spiked into test polymer films at concentrations of 762-1152 mg kg-1 by a solvent soaking technique. Migration (w/w) of surrogate compounds from loaded PP films into Miglyol 812 (a medium-chain triglyceride mixture) and 10% ethanol was quantified by headspace GC/MS during HPP and TP, and subsequent storage at 25°C for up to 10 days. HPP significantly delayed migration of the surrogates from PP into both food simulants relative to TP. The average migrations into Miglyol after TP and HPP were 92.2-109% and 16-60.6%, respectively. Diffusion coefficients estimated by migration modelling showed a reduction of more than two orders of magnitude for all surrogate compounds under high pressure at 700 MPa (AP' = 8.0) relative to equivalent TP at 0.1 MPa (AP' = 13.1). The relative Tg increase of PP copolymer under compression at 700 MPa was estimated as Tg+94°C. For 10% ethanol, average migrations after TP and HPP were 9.3-50.9% and 8.6-22.8%, respectively. During extended storage, migration into both simulants from HPP-treated samples was initially slower than that from untreated or TP-treated films. However, after 8-24 hours of storage, the differences in percent migration of selected surrogates were not significant (p > .05) among the treated PP films. Therefore, the physical changes of PP films that occur during HPP appear to be reversible with a return to their original dimensions and diffusion properties after decompression.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
3-Chlorobenzaldehyde, 97%