Merck
  • Loss of Integrin αvβ8 in Murine Hepatocytes Accelerates Liver Regeneration.
Loss of Integrin αvβ8 in Murine Hepatocytes Accelerates Liver Regeneration.
The American journal of pathology (2018-11-19)
Stephen N Greenhalgh, Kylie P Matchett, Richard S Taylor, Katherine Huang, John T Li, Koy Saeteurn, Mhairi C Donnelly, Eilidh E M Simpson, Joshua L Pollack, Amha Atakilit, Kenneth J Simpson, Jacquelyn J Maher, John P Iredale, Dean Sheppard, Neil C Henderson
ABSTRACT
Recent fate-mapping studies in mice have provided substantial evidence that mature adult hepatocytes are a major source of new hepatocytes after liver injury. In other systems, integrin αvβ8 has a major role in activating transforming growth factor (TGF)-β, a potent inhibitor of hepatocyte proliferation. We hypothesized that depletion of hepatocyte integrin αvβ8 would increase hepatocyte proliferation and accelerate liver regeneration after injury. Using Itgb8flox/flox;Alb-Cre mice to deplete hepatocyte αvβ8, after partial hepatectomy, hepatocyte proliferation and liver-to-body weight ratio were significantly increased in Itgb8flox/flox;Alb-Cre mice compared with control mice. Antibody-mediated blockade of hepatocyte αvβ8 in vitro, with assessment of TGF-β signaling pathways by real-time quantitative PCR array, supported the hypothesis that integrin αvβ8 inhibition alters hepatocyte TGF-β signaling toward a pro-regenerative phenotype. A diethylnitrosamine-induced model of hepatocellular carcinoma, used to examine the possibility that this pro-proliferative phenotype might be oncogenic, revealed no difference in either tumor number or size between Itgb8flox/flox;Alb-Cre and control mice. Immunohistochemistry for integrin αvβ8 in healthy and injured human liver demonstrated that human hepatocytes express integrin αvβ8. Depletion of hepatocyte integrin αvβ8 results in increased hepatocyte proliferation and accelerated liver regeneration after partial hepatectomy in mice. These data demonstrate that targeting integrin αvβ8 may represent a promising therapeutic strategy to drive liver regeneration in patients with a broad range of liver diseases.
Sigma-Aldrich
ECM Cell Adhesion Array Kit, colorimetric, The ECM Cell Adhesion Array kit is useful for assessing specific cell surface integrins, monitoring in vitro cell differentiation, or screening potential cell adhesion promoters/inhibitors.
Sigma-Aldrich
HEPES, ≥99.5% (titration)