Merck
  • Synthesis, antimycobacterial activity evaluation, and QSAR studies of chalcone derivatives.
Synthesis, antimycobacterial activity evaluation, and QSAR studies of chalcone derivatives.
Bioorganic & medicinal chemistry letters (2007-02-06)
P M Sivakumar, S Prabu Seenivasan, Vanaja Kumar, Mukesh Doble
ABSTRACT
In order to develop relatively small molecules as antimycobacterial agents, twenty-five chalcones were synthesized, their activity was evaluated, and quantitative structure-activity relationship (QSAR) was developed. The synthesis was based on the Claisen-Schimdt scheme and the resultant compounds were tested for antitubercular activity by luciferase reporter phage (LRP) assay. Compound C(24) was found to be the most active ( approximately 99%) in this series based on the percentage reduction in Relative Light Units at both 50 and 100 microg/ml levels, followed by compound C(21). Four compounds at the 50 microg/ml and eight compounds at the 100 microg/ml showed activity above 90% level. QSAR model was developed between activity and spatial, topological, and ADME descriptors for the 50 microg/ml data. The statistical measures such as r, r(2), q(2), and F values obtained for the training set were in acceptable range and hence this relationship was used for the test set. The predictive ability of the model is satisfactory (q(2)=0.56) and it can be used for designing similar group of compounds.
Sigma-Aldrich
trans-Chalcone, 97%