Merck
The emerging role of lysophosphatidic acid in cancer.
Nature reviews. Cancer (2003-08-02)
Gordon B Mills, Wouter H Moolenaar
ABSTRACT
The bioactive phospholipid lysophosphatidic acid (LPA) stimulates cell proliferation, migration and survival by acting on its cognate G-protein-coupled receptors. Aberrant LPA production, receptor expression and signalling probably contribute to cancer initiation, progression and metastasis. The recent identification of ecto-enzymes that mediate the production and degradation of LPA, as well as the development of receptor-selective analogues, indicate mechanisms by which LPA production or action could be modulated for cancer therapy.
Product Description
Avanti
17:1 Lyso PA, Avanti Research - A Croda Brand
Pricing and availability is not currently available.