Merck
  • Smooth muscle cells-derived CXCL10 prevents endothelial healing through PI3Kγ-dependent T cells response.
Smooth muscle cells-derived CXCL10 prevents endothelial healing through PI3Kγ-dependent T cells response.
Cardiovascular research (2019-05-21)
Adrien Lupieri, Natalia F Smirnova, Romain Solinhac, Nicole Malet, Mehdi Benamar, Abdel Saoudi, Icia Santos-Zas, Lynda Zeboudj, Hafid Ait-Oufella, Emilio Hirsch, Paul Ohayon, Thibault Lhermusier, Didier Carrié, Jean-François Arnal, Damien Ramel, Stephanie Gayral, Muriel Laffargue
ABSTRACT
Defects in efficient endothelial healing have been associated with complication of atherosclerosis such as post-angioplasty neoatherosclerosis and plaque erosion leading to thrombus formation. However, current preventive strategies do not consider re-endothelialization in their design. Here, we investigate mechanisms linking immune processes and defect in re-endothelialization. We especially evaluate if targeting phosphoinositide 3-kinase γ immune processes could restore endothelial healing and identify immune mediators responsible for these defects. Using in vivo model of endovascular injury, we showed that both ubiquitous genetic inactivation of PI3Kγ and hematopoietic cell-specific PI3Kγ deletion improved re-endothelialization and that CD4+ T-cell population drives this effect. Accordingly, absence of PI3Kγ activity correlates with a decrease in local IFNγ secretion and its downstream interferon-inducible chemokine CXCL10. CXCL10 neutralization promoted re-endothelialization in vivo as the same level than those observed in absence of PI3Kγ suggesting a role of CXCL10 in re-endothelialization defect. Using a new established ex vivo model of carotid re-endothelialization, we showed that blocking CXCL10 restore the IFNγ-induced inhibition of endothelial healing and identify smooth muscle cells as the source of CXCL10 secretion in response to Th1 cytokine. Altogether, these findings expose an unforeseen cellular cross-talk within the arterial wall whereby a PI3Kγ-dependent T-cell response leads to CXCL10 production by smooth muscle cells which in turn inhibits endothelial healing. Therefore, both PI3Kγ and the IFNγ/CXCL10 axis provide novel strategies to promote endothelial healing.
Sigma-Aldrich
Elastase from porcine pancreas, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.