Merck
  • The antibacterial activity of p-tert-butylcalix[6]arene and its effect on a membrane model: molecular dynamics and Langmuir film studies.
The antibacterial activity of p-tert-butylcalix[6]arene and its effect on a membrane model: molecular dynamics and Langmuir film studies.
Physical chemistry chemical physics : PCCP (2020-03-04)
Ellen C Wrobel, Lucas S de Lara, Taiza A S do Carmo, Patrícia Castellen, Márcio Lazzarotto, Sérgio R de Lázaro, Alexandre Camilo, Luciano Caseli, Rolf Schmidt, Christine E DeWolf, Karen Wohnrath
ABSTRACT
The antibacterial activity of a calixarene derivative, p-tert-butylcalix[6]arene (Calix6), was assessed and was shown not to inhibit the growth of E. coli, S. aureus and B. subtilis bacteria. With the aim of gaining more insights into the absence of antibacterial activity of Calix6, the interaction of this derivative with DPPG, a bacterial cell membrane lipid, was studied. Langmuir monolayers were used as the model membrane. Pure DPPG and pure Calix6 monolayers, as well as binary DPPG:Calix6 mixtures were studied using surface pressure measurements, compressional modulus, Brewster angle and fluorescence microscopies, ellipsometry, polarization-modulation infrared reflection absorption spectroscopy and molecular dynamics simulations. Thermodynamic properties of the mixed monolayers were additionally calculated using thermodynamic parameters. The analysis of isotherms showed that Calix6 significantly affects the DPPG monolayers, modifying the isotherm profile and increasing the molecular area, in agreement with the molecular dynamics simulations. The presence of Calix6 in the mixed monolayers decreased the interfacial elasticity, indicating that calixarene disrupts the strong intermolecular interactions of DPPG hindering its organization into a compact arrangement. At low molar ratios of Calix6, the DPPG:Calix6 interactions are preferentially attractive, due to the interactions between the hydrophobic tails of DPPG and the tert-butyl groups of Calix6. Increasing the proportion of calixarene generates repulsive interactions. Calix6 significantly affects the hydrophobic tail organization, which was confirmed by PM-IRRAS measurements. Calix6 appears to be expelled from the mixed films at a biologically relevant surface pressure, π = 30 mN m-1, indicating a low interaction with the cell membrane model related to the absence of antibacterial activity.
Product Description
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131C
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131P, powder
Avanti
16:0 PG, Avanti Research - A Croda Brand