Merck
Biotransformation of xenobiotics by amine oxidases.
Fundamental & clinical pharmacology (2001-07-27)
M S Benedetti
ABSTRACT
Although the cytochrome P450 (CYP) system ranks first in terms of catalytic versatility and the wide range of xenobiotics it detoxifies or activates to reactive intermediates, the contribution of amine oxidases and in particular of monoamine oxidases (MAOs) to the metabolism of xenobiotics is far from negligible but has been largely neglected. In this review on the involvement of amine oxidases in the metabolism of xenobiotics, the major characteristics reported for the CYP system (protein, reaction, tissue distribution, subcellular localisation, substrates, inhibitors, inducers, genetic polymorphism, impact of different physiopathological conditions on the activity, turnover) will be compared, whenever possible, with the corresponding characteristics of amine oxidases (MAOs in particular). The knowledge of the involvement of MAO-A, -B or both in the metabolism of a drug allows us to predict interactions with selective or non-selective MAO inhibitors (e.g. the metabolism of a drug deaminated by both forms of MAO is not necessarily inhibited in vivo by a selective MAO-A or -B inhibitor). If a drug is metabolized by MAOs, competitive interactions can occur with other drugs that are MAO substrates, e.g. with beta-adrenoceptor agonists and antagonists, prodrugs of dopamine, serotonin 5-HT1-receptor agonists as well as with primaquine, flurazepam and citalopram. Moreover, the knowledge of the involvement of MAOs in the metabolism of a drug may suggest possible, although not obligatory, interactions with tyramine-containing food or drink, with over the counter medicines sold to relieve the symptoms of coughs and colds (generally containing the indirectly-acting sympathomimetic amine phenylpropanolamine) or with phenylephrine-containing preparations. Finally, biotransformation by amine oxidases, as by CYP, does not always lead to detoxication but can produce toxic compounds.
Sigma-Aldrich
Monoamine Oxidase A human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Monoamine Oxidase Insect Cell Control, recombinant, expressed in wild-type baculovirus infected BTI insect cells
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells