Merck
  • Impact of consumer behavior on furan and furan-derivative exposure during coffee consumption. A comparison between brewing methods and drinking preferences.
Impact of consumer behavior on furan and furan-derivative exposure during coffee consumption. A comparison between brewing methods and drinking preferences.
Food chemistry (2018-10-13)
Anja Rahn, Chahan Yeretzian
ABSTRACT
This study examined the influence of consumer behavior on furan, 2-methylfuran, 3-methylfuran, 2,5-dimethylfuran and 2,3-dimethylfuran exposure in coffee. Coffees brewed using a filter, fully automatic, capsule machine or reconstituted instant coffee were found to have a significant different cup concentrations of furan derivatives. Coffee brewed with the fully automatic machine contained the highest furan and furan derivative concentrations (99.05 µg/L furan, 263.91 µg/L 2-methylfuran, 13.15 µg/L 3-methylfuran and 8.44 µg/L 2,5-dimethylfuran) whereas soluble coffee did not contain detectable levels, thereby contributing least to a consumer's dietary exposure. Furan and furan derivative concentrations were found to decrease significantly upon cooling, reducing consumer exposure by 8.0-17.2 % on average once the coffee reached drinking temperature 55-60 °C, in ceramic cups. Serving coffee in a ceramic or disposable cup were found to influence the cooling dynamics of the coffee but did not statistically influence the consumers exposure at a given temperature.
Sigma-Aldrich
Furan, ≥99%
Sigma-Aldrich
2,3-Dimethylfuran, 99%
Sigma-Aldrich
3-Methylfuran, AldrichCPR