
Vincristine sulfate

Synonym(s):
Vincristine sulfate salt, 22-Oxovincaleukoblastine sulfate salt, Leurocristine sulfate salt, VCR
Empirical Formula (Hill Notation):
C46H56N4O10 · H2SO4
CAS Number:
Molecular Weight:
923.04
Products
Product Category
Brand
Feature
Formula Weight
Manufacturer
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3