Skip to Content
Merck
Uridine 5′-triphosphate trisodium salt hydrate Type IV, ≥93.0% (HPLC)

Uridine 5′-triphosphate trisodium salt hydrate

Uridine 5′-triphosphate trisodium salt hydrate Type IV, ≥93.0% (HPLC)
Synonym(s):
UTP Na3
Empirical Formula (Hill Notation):
C9H12N2Na3O15P3 · xH2O
CAS Number:
Molecular Weight:
550.09 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Research Area
Quality Segment