Skip to Content
Merck
Trisodium 2-methylcitrate, racemic mixture of diastereomers ≥85% (mixture of diastereomers, NMR)

Trisodium 2-methylcitrate, racemic mixture of diastereomers

Trisodium 2-methylcitrate, racemic mixture of diastereomers ≥85% (mixture of diastereomers, NMR)
Synonym(s):
Trisodium 3-carboxy-3-hydroxy-2-methyl-pentanedioate
Empirical Formula (Hill Notation):
C7H7Na3O7
CAS Number:
Molecular Weight:
272.10

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1