
Tris Buffered Saline

Synonym(s):
TBS, Tris Buffered Saline, with Tween™ 20, pH 8.0
Products
Product Category
Brand
Color
Feature
pH Value
Physical Form
Product Line
Quality Segment
Application
Showing 1-8 of 8
Compare
Product Number
Description
Pricing