Skip to Content
Merck
Trimethylphosphine solution 0.9 M in 2-methyltetrahydrofuran

Trimethylphosphine solution

Trimethylphosphine solution 0.9 M in 2-methyltetrahydrofuran
Synonym(s):
PMe3, Trimethylphosphorus
Linear Formula:
(CH3)3P
CAS Number:
Molecular Weight:
76.08

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type