![Tretinoin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/106/295/5898359c-042b-43bb-ac16-f1c95a917e33/640/5898359c-042b-43bb-ac16-f1c95a917e33.png)
Tretinoin
![Tretinoin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/106/295/5898359c-042b-43bb-ac16-f1c95a917e33/640/5898359c-042b-43bb-ac16-f1c95a917e33.png)
Synonym(s):
Retinoic acid, ATRA, Tretinoin, Vitamin A acid, all-trans-Retinoic acid
Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3