Skip to Content
Merck
Taurolidine >97% (NMR), powder

Taurolidine

Taurolidine >97% (NMR), powder
Synonym(s):
4,4′-Methylenebis(tetrahydro-1,2,4-sulfadiazine 1,1-dioxide), Taurolin, Tauroline
Empirical Formula (Hill Notation):
C7H16N4O4S2
CAS Number:
Molecular Weight:
284.36
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing