
Sumatriptan succinate

Synonym(s):
3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide succinate, GR-43175, Sumatriptan succinate
Empirical Formula (Hill Notation):
C14H21N3O2S · C4H6O4
CAS Number:
Molecular Weight:
413.49
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing