
Alpha Lipoic Acid

Synonym(s):
(±)-α-Lipoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 6,8-Dithiooctanoic acid, DL-α-Lipoic acid, DL-6,8-Thioctic acid, Lip(S2)
Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing