
SP Brij® C2 MBAL-SO-(SG)

Synonym(s):
Brij® 52, Polyethylene glycol hexadecyl ether, Polyoxyethylene (2) cetyl ether
Linear Formula:
C16H33(OCH2CH2)nOH, n~2
CAS Number:
Products
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing