Salbutamol hemisulfate salt
Synonym(s):
α-([t-Butylamino]methyl)-4-hydroxy-m-xylene-α,α′-diol, Albuterol
Linear Formula:
C13H21NO3 · 1/2H2SO4
Products
Product Category
Brand
Formula Weight
Gene Alias
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing