Skip to Content
Merck
(S)-(−)-α,α-Diphenyl-2-pyrrolidinemethanol 99%

(S)-(−)-α,α-Diphenyl-2-pyrrolidinemethanol

(S)-(−)-α,α-Diphenyl-2-pyrrolidinemethanol 99%
Synonym(s):
(S)-(−)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-L-prolinol
Empirical Formula (Hill Notation):
C17H19NO
CAS Number:
Molecular Weight:
253.34

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing