
Racepinephrine Hydrochloride

Synonym(s):
(±)-Epinephrine hydrochloride, (±)-Adrenalin, 4-(1-Hydroxy-2-[methylamino]ethyl)-1,2-benzenediol hydrochloride
Empirical Formula (Hill Notation):
C9H13NO3 · HCl
CAS Number:
Molecular Weight:
219.67
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing