
Prostaglandin F2α tris salt

Synonym(s):
(5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt, PGF2α−Tris
Empirical Formula (Hill Notation):
C20H34O5 · C4H11NO3
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Functional Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing