
Polynucleotide phosphorylase from Synechocystis sp.

Synonym(s):
PNPase, Polyribonucleotide Nucleotidyltransferase
Products
Product Category
Brand
Biological Source
Physical Form
Recombinant Host
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing