Skip to Content
Merck
NovaSeptum® GO Preloaded System with Autoclavable Bag, Vented, 9-port, sterile; β-irradiated

NovaSeptum® GO Preloaded System with Autoclavable Bag, Vented, 9-port,

NovaSeptum® GO Preloaded System with Autoclavable Bag, Vented, 9-port, sterile; β-irradiated
Synonym(s):
NovaSeptum® GO Autoclavable Bag, Vented

Products

Product Category
Brand
Product Line
Agency
Quality Segment
Application
Bottle material
Vial Bottle Material
Sterility
Sterilization Method
Showing 1-1 of 1