
Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide

Linear Formula:
FC6H2(NO2)2NH[CHCH(CH3)2]CONH2
CAS Number:
Molecular Weight:
300.24
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing