![Monoclonal Anti-FITC antibody produced in mouse clone FL-D6, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/292/957/c24c2b1f-42e2-4fdd-966a-5b7cfe24410b/640/c24c2b1f-42e2-4fdd-966a-5b7cfe24410b.jpg)
Monoclonal Anti-FITC antibody produced in mouse
![Monoclonal Anti-FITC antibody produced in mouse clone FL-D6, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/292/957/c24c2b1f-42e2-4fdd-966a-5b7cfe24410b/640/c24c2b1f-42e2-4fdd-966a-5b7cfe24410b.jpg)
Synonym(s):
Monoclonal Anti-FITC
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing