
Methyl ethanesulfonate

Synonym(s):
Ethanesulfonic acid methyl ester
Linear Formula:
C2H5SO3CH3
CAS Number:
Molecular Weight:
124.16
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing