
Mercaptosuccinic acid

Synonym(s):
Thiomalic acid, Mercaptosuccinic acid
Linear Formula:
HOOCCH2CH(SH)COOH
CAS Number:
Molecular Weight:
150.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing