
Mefenpyr-diethyl

Synonym(s):
1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic acid diethyl ester
Empirical Formula (Hill Notation):
C16H18Cl2N2O4
CAS Number:
Molecular Weight:
373.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2