
Ketoprofen Related Compound F

Synonym(s):
2-(3-Benzoylphenyl)propionitrile
Linear Formula:
CH3CH(C6H4COC6H5)CN
CAS Number:
Molecular Weight:
235.28
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2