
Irinotecan Related Compound E

Synonym(s):
(4S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 7-Ethylcamptothecin
Empirical Formula (Hill Notation):
C22H20N2O4
CAS Number:
Molecular Weight:
376.41
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2