
Hydroxyzine pamoate

Synonym(s):
Hydroxyzine pamoate salt
Empirical Formula (Hill Notation):
C21H27ClN2O2 · C23H16O6
CAS Number:
Molecular Weight:
763.27
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2